N-(2-Chlorophenyl)-2-hydrazino-2-oxoacetamide

Supplier: Matrix Scientific

036053-500MG
101840-592EA 290.77 USD
101840-592
N-(2-Chlorophenyl)-2-hydrazino-2-oxoacetamide
N-(2-Chlorophenyl)-2-hydrazino-2-oxoacetamide

Hazard: Irritant

Formula: C₈H₈ClN₃O₂
MDL Number: MFCD02307752
CAS Number: 53117-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR