2-({4-Allyl-5-[(1-naphthylamino)methyl]-4H-1,2,4-triazol-3-yl}thio)acetohydrazide

Supplier: Matrix Scientific

036043-500MG
101840-572EA 387.69 USD
101840-572
2-({4-Allyl-5-[(1-naphthylamino)methyl]-4H-1,2,4-triazol-3-yl}thio)acetohydrazide
2-({4-Allyl-5-[(1-naphthylamino)methyl]-4H-1,2,4-triazol-3-yl}thio)acetohydrazide

Hazard: Irritant

Formula: C₁₈H₂₀N₆OS
MDL Number: MFCD02255655
CAS Number: 1071368-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR