2-Chloro-5-(trifluoromethyl)benzamide

Supplier: Matrix Scientific

002648-5G 002648-1G
101784-388EA 47.69 USD
101784-388 101784-390
2-Chloro-5-(trifluoromethyl)benzamide
2-Chloro-5-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₈H₅ClF₃NO
Melting Pt: 142…144 °C
MDL Number: MFCD01631467
CAS Number: 20566-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR