2-Chloro-4,5-difluorobenzenesulfonamide

Supplier: Matrix Scientific

002417-1G
101783-874EA 83.08 USD
101783-874
2-Chloro-4,5-difluorobenzenesulfonamide
2-Chloro-4,5-difluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₄ClF₂NO₂S
MDL Number: MFCD02091389
CAS Number: 287172-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR