2-Chloro-4,5-difluorobenzenesulfonyl chloride
Supplier: Matrix Scientific
002416-1G
101783-872EA
58.46
USD
101783-872
2-Chloro-4,5-difluorobenzenesulfonyl chloride
2-Chloro-4,5-difluorobenzenesulphonyl chloride
Hazard: Irritant
Formula:
C₆H₂Cl₂F₂O₂S |
MDL Number:
MFCD02091388 CAS Number: 67475-58-5 UN: 1760 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...