Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-(trifluoromethyl)benzyl alcohol

Supplier: Matrix Scientific


002306-5G 002306-1G
101783-656EA 158.23 USD InStock
101783-656 101783-654
2-Chloro-3-(trifluoromethyl)benzyl alcohol
2-Chloro-3-(trifluoromethyl)benzyl alcohol
CAS: 261763-20-6
MDL Number: MFCD01631537
Molecular Formula: C8H6ClF3O
Molecular Weight: 210,58
Hazard: Irritant


Formula: C₈H₆ClF₃O
MW: 210,58
MDL Number: MFCD01631537
CAS Number: 261763-20-6
EINECS: 000-000-0
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty