3-Chloro-2-fluorobenzamide

Supplier: Matrix Scientific

002091-5G 002091-1G
101783-188EA 38.46 USD
101783-188 101783-190
3-Chloro-2-fluorobenzamide
3-Chloro-2-fluorobenzamide

Hazard: Irritant

Formula: C₇H₅ClFNO
MDL Number: MFCD00042205
CAS Number: 104326-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR