3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde

Supplier: Matrix Scientific

002072-5G
101783-154EA 141.54 USD
101783-154
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde
3-Chloro-2-fluoro-6-(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₈H₃ClF₄O
MDL Number: MFCD01631343
CAS Number: 186517-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR