3-Fluoro-2-(trifluoromethyl)benzamide ≥97%

Supplier: Matrix Scientific

002008-1G
101783-052EA 283.08 USD
101783-052
3-Fluoro-2-(trifluoromethyl)benzamide ≥97%
3-Fluoro-2-(trifluoromethyl)benzamide

Hazard: Irritant

Formula: C₈H₅F₄NO
MDL Number: MFCD01631492
CAS Number: 287398-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR