4-Chloro-2,5-difluorobenzenesulfonamide

Supplier: Matrix Scientific

001956-1G
101782-928EA 60 USD
101782-928
4-Chloro-2,5-difluorobenzenesulfonamide
4-Chloro-2,5-difluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₄ClF₂NO₂S
Melting Pt: 121…122 °C
MDL Number: MFCD02091387
CAS Number: 287172-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR