4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

001523-1G
101781-880EA 53.85 USD
101781-880
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid ≥97%
4-Hydroxy-8-(trifluoromethyl)quinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃NO₃
Melting Pt: 292 °C
MDL Number: MFCD00047603
CAS Number: 23779-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR