5-Formyl-2-methoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Formyl-2-methoxybenzeneboronic acid(5-Formyl-2-methoxyphenyl)boronic acid

001505-1G
101781-840EA 15.38 USD
101781-840
5-Formyl-2-methoxyphenylboronic acid ≥97%
5-Formyl-2-methoxyphenylboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₈H₉BO₄
MW: 179.9700 g/mol
Melting Pt: 160 °C
MDL Number: MFCD01319044
CAS Number: 127972-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR