5-Bromo-2,4-dimethoxypyrimidine ≥98%
Supplier: Matrix Scientific
001248-25G
001248-5G
101781-244EA
49.23
USD
101781-244
101781-246
5-Bromo-2,4-dimethoxypyrimidine ≥98%
5-Bromo-2,4-dimethoxypyrimidine
Hazard: Irritant/Light Sensitive/Refrigerate
Formula:
C₆H₇BrN₂O₂ MW: 219.0400 g/mol Melting Pt: 62…65 °C |
MDL Number:
MFCD00038016 CAS Number: 56686-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...