5-Bromo-2,4-difluorobenzenesulfonamide

Supplier: Matrix Scientific

001244-5G 001244-1G
101781-232EA 49.23 USD
101781-232 101781-234
5-Bromo-2,4-difluorobenzenesulfonamide
5-Bromo-2,4-difluorobenzenesulphonamide

Hazard: Irritant

Formula: C₆H₄BrF₂NO₂S
MDL Number: MFCD01940425
CAS Number: 287172-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR