MATR001219

001219-5G
101781-186EA 81.08 USD
101781-186
MATR001219
Purity: 97%
CAS: 2264-01-9
MDL Number: MFCD00078412
Molecular Formula: C12H10F8O4
Molecular Weight: 370,2
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR