(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride

Supplier: Matrix Scientific

001214-100MG
101781-170EA 103.08 USD
101781-170
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride

Hazard: Irritant

Formula: C₄H₇ClF₃NO₂
Melting Pt: 168…170 °C
MDL Number: MFCD00181951
CAS Number: 91291-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR