3-(2-Fluorophenoxy)benzyl bromide

Supplier: Matrix Scientific

Synonyms: 3'-Bromomethyl-2-fluorodiphenyl Ether

006063-100MG
101789-990EA 112.31 USD
101789-990
3-(2-Fluorophenoxy)benzyl bromide
3-(2-Fluorophenoxy)benzyl bromide

Hazard: Irritant

Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Density: 1.46
MDL Number: MFCD00059914
CAS Number: 242812-04-0

Order Now

Specification Test Results

Density 1.46

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR