2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: Matrix Scientific

008879-5G
101795-700EA 273.85 USD
101795-700
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane ≥97%
2-(3-Chloropropyl)-2-(4-fluorophenyl)-1,3-dioxolane

Hazard: Irritant

Formula: C₁₂H₁₄ClFO₂
MW: 244.69 g/mol
Boiling Pt: 101…103 °C (0.2 Torr)
Density: 1.219
MDL Number: MFCD00020909
CAS Number: 3308-94-9

Order Now

Specification Test Results

Density 1.219
ND20 1.506

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR