3-Phenylbenzyl isothiocyanate

Supplier: Matrix Scientific

057178-5G
101912-654EA 969.23 USD
101912-654
3-Phenylbenzyl isothiocyanate
3-Phenylbenzyl isothiocyanate

Hazard: Irritant

Formula: C₁₄H₁₁NS
Boiling Pt: 165…170 °C (1 mmHg)
MDL Number: MFCD09025720
CAS Number: 1001569-47-6

Order Now

Specification Test Results

Boiling point 165-170°C/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR