Methyl-3-hydroxy-4-methoxyphenylacetate

Supplier: Matrix Scientific

057140-25G
101912-514EA 961.54 USD
101912-514
Methyl-3-hydroxy-4-methoxyphenylacetate
Methyl-3-hydroxy-4-methoxyphenylacetate

Hazard: Irritant

Formula: C₁₀H₁₂O₄
Boiling Pt: 131 °C (1 mmHg)
MDL Number: MFCD17168381
CAS Number: 15964-81-5

Order Now

Specification Test Results

Boiling point 131°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR