Ethyl-2-nitrobuytrate ≥96%

Supplier: Matrix Scientific

057075-5G 057075-1G
101912-274EA 513.85 USD
101912-274 101912-272
Ethyl-2-nitrobuytrate ≥96%
Ethyl-2-nitrobuytrate

Hazard: Irritant

Formula: C₆H₁₁NO₄
Boiling Pt: 82…83 °C (8 mmHg)
MDL Number: MFCD00075585
CAS Number: 2531-81-9

Order Now

Specification Test Results

Boiling point 82-83°C/8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR