Ethyl-3-chloropicolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-chloro-2-pyridinecarboxylate
, Ethyl 3-chloropicolinate
048691-500MG
048691-1G
101884-888EA
318.46
USD
101884-888
101884-890
Ethyl-3-chloropicolinate ≥95%
Ethyl 3-chloropicolinate
Hazard: Irritant
Formula:
C₈H₈ClNO₂ MW: 185.6076 g/mol Boiling Pt: 104…106 °C |
MDL Number:
MFCD11656307 CAS Number: 128073-20-1 |
Specification Test Results
Boiling point | 104-106@0. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...