2-Nitro-5-(trifluoromethyl)benzenol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Hydroxy-4-nitrobenzotrifluoride2-Nitro-5-(trifluoromethyl)phenol

048213-500MG 048213-1G
101883-318EA 128.8 USD
101883-318 101883-320
2-Nitro-5-(trifluoromethyl)benzenol ≥95%
2-Nitro-5-(trifluoromethyl)benzenol

Hazard: Irritant

Formula: C₇H₄F₃NO₃
MW: 207.11 g/mol
Boiling Pt: 108…111 °C (34 mmHg)
MDL Number: MFCD08282785
CAS Number: 402-17-5

Order Now

Specification Test Results

Boiling point 108-111°C@3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR