N-Benzyl-N-(3-butenyl)amine ≥95%

Supplier: Matrix Scientific

048032-1G
101882-474EA 182.85 USD
101882-474
N-Benzyl-N-(3-butenyl)amine ≥95%
N-Benzyl-N-(3-butenyl)amine

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 65 °C (1 mmHg)
MDL Number: MFCD01249643
CAS Number: 17150-62-8

Order Now

Specification Test Results

Boiling point 65°C/1mm°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR