1-Cyclohexyltetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

047916-1G 047916-500MG 047916-5G
101881-990EA 602.6 USD
101881-990 101881-986 101881-988
1-Cyclohexyltetrahydro-4(1H)-pyridinone ≥95%
1-Cyclohexyltetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₁H₁₉NO
Boiling Pt: 95 °C (0.4 mmHg)
MDL Number: MFCD03840833
CAS Number: 16771-84-9

Order Now

Specification Test Results

Boiling point 95°C/0.4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR