1-Hexyl-4-nitro-1H-pyrazole ≥95%
Supplier: Matrix Scientific
047809-500MG
047809-1G
101881-540EA
467.69
USD
101881-540
101881-538
1-Hexyl-4-nitro-1H-pyrazole ≥95%
1-Hexyl-4-nitro-1H-pyrazole
Hazard: Irritant
Formula:
C₉H₁₅N₃O₂ Boiling Pt: 170 °C (0.1 mmHg) |
MDL Number:
MFCD11100253 CAS Number: 1171685-13-4 |
Specification Test Results
Boiling point | 170°C/0.1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...