3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonan-9-one ≥95%
Supplier: Matrix Scientific
047773-5G
047773-500MG
047773-1G
101881-362EA
500
USD
101881-362
101881-364
101881-366
3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonan-9-one ≥95%
3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonan-9-one
Hazard: Irritant
Formula:
C₁₅H₂₈N₂O Boiling Pt: 120 °C (0.15 mmHg) |
MDL Number:
MFCD09027160 CAS Number: 85928-06-9 |
Specification Test Results
Boiling point | 120°C/0.15m |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...