1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine ≥95%
Supplier: Matrix Scientific
045627-5G
045627-1G
045627-500MG
101871-820EA
923.08
USD
101871-820
101871-816
101871-818
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]piperidine
Hazard: Irritant
Formula:
C₉H₁₃ClN₂S Boiling Pt: 102 °C (0.1 mmHg) |
MDL Number:
MFCD01443899 CAS Number: 339104-71-1 |
Specification Test Results
Boiling point | 102°C/0.1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...