Methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%

Supplier: Matrix Scientific

043207-5G 043207-1G 043207-500MG
101859-680EA 181.54 USD
101859-680 101859-684 101859-682
Methyl-2-(1H-1,2,4-triazol-1-yl)propanoate ≥95%
Methyl-2-(1H-1,2,4-triazol-1-yl)propanoate

Hazard: Irritant

Formula: C₆H₉N₃O₂
Boiling Pt: 90 °C (0.5 mmHg)
MDL Number: MFCD00215246
CAS Number: 100554-35-6

Order Now

Specification Test Results

Boiling point 90°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR