Diethyl-2-[(4-methoxyphenyl)methylene]malonate ≥95%

Supplier: Matrix Scientific

043035-1G
101858-826EA 262.2 USD
101858-826
Diethyl-2-[(4-methoxyphenyl)methylene]malonate ≥95%
Diethyl-2-[(4-methoxyphenyl)methylene]malonate

Hazard: Irritant

Formula: C₁₅H₁₈O₅
Boiling Pt: 156…158 °C (0.1 mmHg)
MDL Number: MFCD00182557
CAS Number: 6768-23-6

Order Now

Specification Test Results

Boiling point 156-158°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR