Ethyl-2-(2-chloro-6-fluorobenzyl)-3-oxobutanoate ≥95%
Supplier: Matrix Scientific
043024-1G
101858-766EA
473.85
USD
101858-766
Ethyl-2-(2-chloro-6-fluorobenzyl)-3-oxobutanoate ≥95%
Ethyl-2-(2-chloro-6-fluorobenzyl)-3-oxobutanoate
Hazard: Irritant
Formula:
C₁₃H₁₄ClFO₃ Boiling Pt: 110 °C (0.1 mmHg) |
MDL Number:
MFCD00052428 CAS Number: 477856-17-0 |
Specification Test Results
Boiling point | 110°C/0.1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...