3-Bromo-2-(trifluoromethyl)pyridine ≥97%

Supplier: Matrix Scientific

032388-1G
101833-620EA 56.92 USD
101833-620
3-Bromo-2-(trifluoromethyl)pyridine ≥97%
3-Bromo-2-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃BrF₃N
MW: 225.9939 g/mol
Boiling Pt: 84…86 °C (20 mmHg)
MDL Number: MFCD08458124
CAS Number: 590371-58-7

Order Now

Specification Test Results

Boiling point 84-86°C/20m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR