2-Bromo-3-fluoropropionic acid ≥97%

Supplier: Matrix Scientific

024676-250MG
101818-620EA 138.46 USD
101818-620
2-Bromo-3-fluoropropionic acid ≥97%
2-Bromo-3-fluoropropionic acid

Hazard: Irritant

Formula: C₃H₄BrFO₂
Boiling Pt: 62…63 °C (0.2 mmHg)
MDL Number: MFCD09800640
CAS Number: 39621-37-9

Order Now

Specification Test Results

Boiling point 62-63°C/0.2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR