3,3-Dimethylpiperidine ≥95%

Supplier: Matrix Scientific

012902-1G
101803-592EA 368 USD
101803-592
3,3-Dimethylpiperidine ≥95%
3,3-Dimethylpiperidine

Hazard: Irritant

Formula: C₇H₁₅N
Boiling Pt: 134…136 °C
MDL Number: MFCD00005995
CAS Number: 1193-12-0

Order Now

Specification Test Results

Boiling point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR