Perfluoroacetophenone ≥98%

Supplier: Matrix Scientific

012867-5G
101803-542EA 423.08 USD
101803-542
Perfluoroacetophenone ≥98%
Perfluoroacetophenone

Hazard: Irritant

Formula: C₈F₈O
Boiling Pt: 130…131 °C
MDL Number: MFCD00216596
CAS Number: 652-22-2

Order Now

Specification Test Results

Boiling point 130-131°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR