Ethyl-7-bromoheptanoate ≥96%
Supplier: Matrix Scientific
Synonyms:
Ethyl 7-bromoheptanoate
, 7-Bromoheptanoic Acid Ethyl Ester
007590-100G
007590-25G
101793-070EA
615.38
USD
101793-070
101793-068
Ethyl-7-bromoheptanoate ≥96%
Ethyl-7-bromoheptanoate
Purity: 96%
CAS: 29823-18-5
MDL Number: MFCD00037103
Molecular Formula: C9H17BrO2
Molecular Weight: 237,14
Boiling Point: 90°C/ .1mm
Hazard: Irritant
Formula:
C₉H₁₇BrO₂ MW: 237.14 g/mol Boiling Pt: 90 °C ( .1 mmHg) |
MDL Number:
MFCD00037103 CAS Number: 29823-18-5 |
Specification Test Results
Boiling point | 90°C/ .1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...