Ethyl-2-bromohexanoate ≥97%
Supplier: Matrix Scientific
007589-5G
007589-25G
101793-066EA
114.65
USD
101793-066
101793-064
Ethyl-2-bromohexanoate ≥97%
Ethyl 2-bromohexanoate
Hazard: Irritant
Formula:
C₈H₁₅BrO₂ Boiling Pt: 213…215 °C |
UN:
3265 ADR: 8,III |
Specification Test Results
Boiling point | 213°C-215°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...