2-Bromodiethylacetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-2-ethylbutanoic acid

007517-25G 007517-5G
101792-968EA 792.31 USD
101792-968 101792-966
2-Bromodiethylacetic acid ≥98%
2-Bromodiethylacetic acid

Hazard: Irritant

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
Boiling Pt: 120 °C (75 mmHg)
MDL Number: MFCD00046512
CAS Number: 5456-23-5

Order Now

Specification Test Results

Boiling point 120°C/75mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR