2-Aminononadecane ≥98%

Supplier: Matrix Scientific

007501-5G
101792-940EA 95.38 USD
101792-940
2-Aminononadecane ≥98%
2-Aminononadecane

Hazard: Irritant

Formula: C₁₉H₄₁N
Boiling Pt: 151 °C (1 mmHg)
MDL Number: MFCD00053641
CAS Number: 31604-55-4

Order Now

Specification Test Results

Boiling point 151°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR