Bis(2,2,2-trifluoroethyl)maleate ≥97%
Supplier: Matrix Scientific
Synonyms:
Bis(2,2,2-trifluoroethyl) (2Z)-but-2-ene-1,4-dioate
, Maleic acid bis(2,2,2-trifluoroethyl)ester
007163-5G
007163-25G
101792-114EA
381.54
USD
101792-114
101792-112
Bis(2,2,2-trifluoroethyl)maleate ≥97%
Bis(2,2,2-trifluoroethyl)maleate
Hazard: Irritant
Formula:
C₈H₆F₆O₄ MW: 280.121 g/mol Boiling Pt: 50 °C (144 mmHg) |
MDL Number:
MFCD00078313 CAS Number: 156631-37-7 |
Specification Test Results
Boiling point | 50°C/144mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...