(S)-(-)-1,1,1-Trifluorodecan-2-ol ≥ ee 98%

Supplier: Matrix Scientific

007102-1G 007102-5G
101792-030EA 467.69 USD
101792-030 101792-028
(S)-(-)-1,1,1-Trifluorodecan-2-ol ≥ ee 98%
(S)-(-)-1,1,1-Trifluorodecan-2-ol

Hazard: Irritant

Formula: C₁₀H₁₉F₃O
Boiling Pt: 66…67 °C (4 mmHg)
MDL Number: MFCD06799354
CAS Number: 111423-27-9

Order Now

Specification Test Results

Boiling point 66-67°C/4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR