2-Bromotetrafluoropropionic acid ≥97%

006358-25G
101790-412EA 299 USD
101790-412
2-Bromotetrafluoropropionic acid ≥97%
2-Bromotetrafluoropropionic acid

Hazard: Corrosive/Irritant

Formula: C₃HBrF₄O₂
MW: 224.94 g/mol
Boiling Pt: 58…60 °C (22 mmHg)
MDL Number: MFCD02183518
CAS Number: 13859-31-9

Order Now

Specification Test Results

Boiling point 58-60°C/22m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR