Dimethyl tetrafluorosuccinate ≥98%
Supplier: Matrix Scientific
Synonyms:
Tetrafluorosuccinic Acid Dimethyl Ester
, Dimethyl Perfluorosuccinate
, Dimethyl Tetrafluorobutanedioate
, Perfluorosuccinic Acid Dimethyl Ester
, Tetrafluorobutanedioic Acid Dimethyl Ester
006283-25G
006283-5G
006283-100G
101790-326EA
33.85
USD
101790-326
101790-328
101790-330
Dimethyl tetrafluorosuccinate ≥98%
Dimethyl tetrafluorosuccinate
Purity: 98%
CAS: 356-36-5
MDL Number: MFCD00043947
Molecular Formula: C6H6F4O4
Molecular Weight: 218,11
Boiling Point: 177°C
Hazard: Irritant
Formula:
C₆H₆F₄O₄ MW: 218.10 g/mol Boiling Pt: 177 °C |
MDL Number:
MFCD00043947 CAS Number: 356-36-5 |
Specification Test Results
Boiling point | 177°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...