(4-Fluorophenylthio)acetone ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Fluorophenylthio)propan-2-one

005731-1G 005731-5G
101789-456EA 412.31 USD
101789-456 101789-454
(4-Fluorophenylthio)acetone ≥97%
(4-Fluorophenylthio)acetone

Hazard: Irritant

Formula: C₉H₉FOS
MW: 184.23
Boiling Pt: 100…103 °C (1 mmHg)
MDL Number: MFCD00179340
CAS Number: 2968-13-0

Order Now

Specification Test Results

Boiling point 100-103°C/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR