5-Bromo-2-butylbenzoxazole ≥95%

Supplier: Matrix Scientific

047388-1G
101879-774EA 467.69 USD
101879-774
5-Bromo-2-butylbenzoxazole ≥95%
5-Bromo-2-butylbenzoxazole

Hazard: Irritant

Formula: C₁₁H₁₂BrNO
Boiling Pt: 125 °C (0.5 mmHg)
MDL Number: MFCD11100251
CAS Number: 1048914-34-6

Order Now

Specification Test Results

Boiling point 125°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR