Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethylpyrazolo[1,5-a]pyridine-3-carboxylate

023803-1G
101816-790EA 85.1 USD
101816-790
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate ≥95%
Ethyl pyrazolo[1,5-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.1986 g/mol
Boiling Pt: 134…135 °C (3 mmHg)
MDL Number: MFCD06411542
CAS Number: 16205-44-0

Order Now

Specification Test Results

Boiling point 134-135°C/3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR