(S)-1-(2-Fluorophenyl)ethylamine ≥97%, ee 98%

Supplier: Matrix Scientific

Synonyms: (S)-1-(2-Fluorophenyl)ethanamine

023715-250MG 023715-1G
101816-556EA 98.46 USD
101816-556 101816-558
(S)-1-(2-Fluorophenyl)ethylamine ≥97%, ee 98%
(S)-1-(2-Fluorophenyl)ethylamine

Hazard: Irritant

Formula: C₈H₁₀FN
MW: 139.17 g/mol
Boiling Pt: 62…63 °C (6 mmHg)
MDL Number: MFCD03092994
CAS Number: 68285-25-6

Order Now

Specification Test Results

Boiling point 62-63°C/6mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR