5-Bromo-2-(trifluoromethyl)pyridine ≥98%
Supplier: Matrix Scientific
018137-25G
101805-490EA
175.38
USD
101805-490
5-Bromo-2-(trifluoromethyl)pyridine ≥98%
5-Bromo-2-(trifluoromethyl)pyridine
Hazard: Irritant
Formula:
C₆H₃BrF₃N MW: 226.00 g/mol Boiling Pt: 68…70 °C (10 mmHg) |
MDL Number:
MFCD06657686 CAS Number: 436799-32-5 |
Specification Test Results
Boiling point | 68-70°C/10m |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...