2,2-Difluorocyclopropanecarboxylic acid ≥95%

Supplier: Matrix Scientific

009429-1G
101797-082EA 106.95 USD
101797-082
2,2-Difluorocyclopropanecarboxylic acid ≥95%
2,2-Difluorocyclopropanecarboxylic acid

Hazard: Irritant

Formula: C₄H₄F₂O₂
MW: 122.07 g/mol
Boiling Pt: 70 °C (7 mmHg)
Melting Pt: 61…63 °C
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Boiling point 70°C/7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR