(2-Chloro-1,3-thiazol-5-yl)methyl acetate ≥95%
Supplier: Matrix Scientific
045338-500MG
045338-1G
045338-5G
101870-392EA
556.92
USD
101870-392
101870-396
101870-394
(2-Chloro-1,3-thiazol-5-yl)methyl acetate ≥95%
(2-Chloro-1,3-thiazol-5-yl)methyl acetate
Hazard: Irritant
Formula:
C₆H₆NO₂ClS Boiling Pt: 75…80 °C (1 mmHg) |
MDL Number:
MFCD01443869 CAS Number: 339018-65-4 |
Specification Test Results
Boiling point | 75-80°C/1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...